Check Order Status
Sodium Chloride (Granular/USP/FCC), Fisher Chemical™
Element Search
Structure Search
Sodium Chloride (Granular/USP/FCC), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMS640-3
3 kg
Poly Bottle
Catalog number
FCMS640-3
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
3 kg
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point
1461°C
Chemical Name or Material
Sodium Chloride
Melting Point
801°C
CAS
7647-14-5
ChemAlert Storage Symbol
Gray
View more