Check Order Status
Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Lauryl Sulfate (Powder/NF/FCC), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
3 kg
Packaging:
View all
Glass Bottle
Catalog number
FCMS529-500
Price (MYR)
615.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
615.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium Lauryl Sulfate
Color
White
CAS
151-21-3
ChemAlert Storage Symbol
Red
Physical Form
Solid
View more