Check Order Status
Starch, Soluble (Powder/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Starch, Soluble (Powder/Certified ACS), Fisher Chemical™
For iodometry
Have Questions?
Catalog Number
Quantity
Packaging
FCMS516-100
100 g
Glass Bottle
Catalog number
FCMS516-100
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Starch, Soluble
Color
White
pH
5.0 to 7.0
CAS
9005-84-9
ChemAlert Storage Symbol
Gray
View more