Check Order Status
Sodium Phosphate Monobasic Monohydrate (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Phosphate Monobasic Monohydrate (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMS369-1
1 kg
Poly Bottle
Catalog number
FCMS369-1
Price (MYR)
525.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
525.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium Phosphate Monobasic Monohydrate
Melting Point
100°C
CAS
10049-21-5
ChemAlert Storage Symbol
Gray
Color
White
View more