Check Order Status
Sodium Iodide (Crystalline/Certified), Fisher Chemical™
Element Search
Structure Search
Sodium Iodide (Crystalline/Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMS324-100
100 g
Glass Bottle
Catalog number
FCMS324-100
Price (MYR)
546.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
546.00
EA
Add to cart
Specifications
Boiling Point
1300°C
Chemical Name or Material
Sodium Iodide
Melting Point
661°C
CAS
7681-82-5
ChemAlert Storage Symbol
Gray
View more