Check Order Status
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide (Pellets/Certified ACS), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
100 g
1 kg
3 kg
10 kg
Packaging:
View all
Poly Bottle
Catalog number
FCMS318-500
Price (MYR)
426.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
426.00
EA
Add to cart
Specifications
Chemical Name or Material
Sodium Hydroxide
pH
14
Cleanup Symbol
Caustic
Color
White
Boiling Point
1390°C
View more