Check Order Status
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMS311-500
500 g
Poly Bottle
Catalog number
FCMS311-500
Price (MYR)
542.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
542.00
BOT
Add to cart
Specifications
Chemical Name or Material
Ethylenediamine Tetraacetic Acid, Disodium Salt Dihydrate
CAS
6381-92-6
ChemAlert Storage Symbol
Gray
Health Hazard 2
CAUTION!
Decomposition Information
>252°C
View more