Check Order Status
Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Potassium Chloride (Crystalline/Certified ACS), Fisher Chemical™
Potassium chloride for calomel cells
Have Questions?
Catalog Number
Quantity
Packaging
FCMP217-500
500 g
Poly Bottle
Catalog number
FCMP217-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Boiling Point
1420°C
Chemical Name or Material
Potassium Chloride
Melting Point
770°C
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
View more