Check Order Status
Isooctane (Pesticide), Fisher Chemical
Element Search
Structure Search
Isooctane (Pesticide), Fisher Chemical
Packaged under nitrogen
Have Questions?
Catalog Number
Quantity
Packaging
FCMO297-4
4 L
Glass Bottle
Catalog number
FCMO297-4
Price (MYR)
1,031.00
EA
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,031.00
EA
Add to cart
Specifications
Chemical Name or Material
Isooctane
Cleanup Symbol
Solvent
Melting Point
-107°C
CAS
540-84-1
ChemAlert Storage Symbol
Red
View more