Check Order Status
Hydroxynaphthol Blue (Certified), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Hydroxynaphthol Blue (Certified), Fisher Chemical
Used as an indicator for EDTA titrimetric determination of calcium. Color changes from reddish pink to blue
Have Questions?
Catalog Number
Quantity
FCMH346-100
100 g
Catalog number
FCMH346-100
Price (MYR)
813.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
813.00
EA
Add to cart
Specifications
Chemical Name or Material
Hydroxynaphthol Blue
CAS
63451-35-4
ChemAlert Storage Symbol
Gray
Molecular Formula
C20H11N2Na3O11S3
MDL Number
MFCD00036393
View more