Check Order Status
Hematoxylin (Certified Biological Stain), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Hematoxylin (Certified Biological Stain), Fisher Chemical™
For use in histology and cytology
Have Questions?
Catalog Number
Quantity
Packaging
FCMH345-25
25 g
Glass Bottle
Catalog number
FCMH345-25
Price (MYR)
913.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
913.00
EA
Add to cart
Specifications
Chemical Name or Material
Hematoxylin
Melting Point
100°C
CAS
517-28-2
ChemAlert Storage Symbol
Gray
Bio. Stain Comm. Certification
Pass Test
View more