Check Order Status
Hydrogen Peroxide, 50% (Certified), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
Hydrogen Peroxide, 50% (Certified), Fisher Chemical
Have Questions?
Catalog Number
Quantity
FCMH341-500
500 mL
Catalog number
FCMH341-500
Price (MYR)
429.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
429.00
EA
Add to cart
Specifications
Chemical Name or Material
Hydrogen Peroxide, 50%
Color
Colorless
Name Note
Stabilized
CAS
7722-84-1
ChemAlert Storage Symbol
Yellow
View more