Hydrogen Peroxide, Fisher Chemical™
Hydrogen Peroxide, Fisher Chemical™
Hydrogen Peroxide, Fisher Chemical™
Hydrogen Peroxide, Fisher Chemical™
Fisher Chemical
Hydrogen Peroxide, Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
FCMH312P-44 LPoly Bottle
Catalog number FCMH312P-4
Price (MYR)
491.00
EA
Add to cart
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
491.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydrogen Peroxide
ColorUndesignated
Name Note3%
AcidityPass Test
CAS7722-84-1
View more