Hexanes, Optima™ for HPLC and GC, Fisher Chemical™
Hexanes, Optima™ for HPLC and GC, Fisher Chemical™
Hexanes, Optima™ for HPLC and GC, Fisher Chemical™
Hexanes, Optima™ for HPLC and GC, Fisher Chemical™
Hexanes, Optima™ for HPLC and GC, Fisher Chemical™
Fisher Chemical
Hexanes, Optima™ for HPLC and GC, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, GC/MS, and pesticide residue analysis | CAS: 92112-69-1 | C6H14 | 86.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Packaging:
View all
Amber Soda Lime Glass
Catalog number FCMH303-4
Price (MYR)
680.00
BOT
Add to cart
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Price (MYR)
680.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHexanes
Cleanup SymbolSolvent
Melting Point-95°C
Absorbance1 AU max. at 195nm, 0.25 AU max. at 210nm, 0.07 AU max. at 220nm, 0.005 AU max. at 254nm
CAS92112-69-1
View more
High purity and versatile solvent suitable for HPLC, spectrophotometry, GC/MS, and pesticide residue analysis
  • Mixture of hexanes where n-hexane content is ≥60%
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis and tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only