Glycerol (Certified ACS), Fisher Chemical™
Glycerol (Certified ACS), Fisher Chemical™
Glycerol (Certified ACS), Fisher Chemical™
Glycerol (Certified ACS), Fisher Chemical™
Glycerol (Certified ACS), Fisher Chemical™
Glycerol (Certified ACS), Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
FCMG33-11 LHDPE Bottle
Catalog number FCMG33-1
Price (MYR)
461.00
EA
Add to cart
Quantity:
1 L
Packaging:
HDPE Bottle
Request bulk or custom format
Price (MYR)
461.00
EA
Add to cart
Specifications
Chemical Name or MaterialGlycerol
CAS56-81-5
ChemAlert Storage SymbolGray
Health Hazard 2WARNING!
Decomposition Information>290°C
View more