Check Order Status
Glutaraldehyde Solution, 50% (Certified), Fisher Chemical
Element Search
Structure Search
Glutaraldehyde Solution, 50% (Certified), Fisher Chemical
Have Questions?
Catalog Number
Quantity
FCMG151-1
1 L
Catalog number
FCMG151-1
Price (MYR)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material
Glutaraldehyde Solution, 50%
Physical Form
Liquid
Melting Point
-33°C
Boiling Point
101.5°C
CAS
111-30-8
View more