Check Order Status
Ethyl Acetate, Optima™ for HPLC and GC, Thermo Scientific™
Element Search
Structure Search
Fisher Chemical
Ethyl Acetate, Optima™ for HPLC and GC, Thermo Scientific™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Catalog Number
Quantity
Packaging
FCME196-4
4 L
Amber Soda Lime Glass
Catalog number
FCME196-4
Price (MYR)
773.00
EA
Add to cart
Quantity:
4 L
Packaging:
Amber Soda Lime Glass
Request bulk or custom format
Price (MYR)
773.00
EA
Add to cart
Specifications
Chemical Name or Material
Ethyl Acetate
Cleanup Symbol
Solvent
Melting Point
-83.5°C
Absorbance
0.015 AU max. at 270nm, 0.12 AU max. at 260nm, 1.00 AU max. at 255nm
CAS
141-78-6
View more
Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
Also meets ACS specifications
Intended for research use only