Check Order Status
1,2-Dichloroethane (Certified ACS), Fisher Chemical™
Element Search
Structure Search
1,2-Dichloroethane (Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCME175-20
20 L
Steel Pail
Catalog number
FCME175-20
Price (MYR)
-
Quantity:
20 L
Packaging:
Steel Pail
Request bulk or custom format
Specifications
Chemical Name or Material
1, 2-Dichloroethane
Cleanup Symbol
Solvent
Melting Point
-35°C
CAS
107-06-2
ChemAlert Storage Symbol
Red
View more