Methylene Chloride (HPLC), Fisher Chemical™
Methylene Chloride (HPLC), Fisher Chemical™
Methylene Chloride (HPLC), Fisher Chemical™
Methylene Chloride (HPLC), Fisher Chemical™
Methylene Chloride (HPLC), Fisher Chemical™
Methylene Chloride (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations.
Have Questions?
Catalog NumberQuantityPackaging
FCMD143-44 LGlass Bottle
Catalog number FCMD143-4
Price (MYR)
597.00
EA
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
597.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethylene Chloride
Cleanup SymbolSolvent
Melting Point-97°C
Name NoteStabilized
Absorbance0.01 AU max., 0.01 AU max. at 260nm, 0.014 AU max. at 254nm, 0.10 max. at 250nm, 0.2 AU max. at 240nm, 1 AU max. at 235nm, Pass Test
View more
Contains approx. 15-200 ppm preservative