Dimethyl Sulfoxide DMSO GC, Fisher Chemical™
Dimethyl Sulfoxide DMSO GC, Fisher Chemical™
Actual product may vary
Dimethyl Sulfoxide DMSO GC, Fisher Chemical™
Dimethyl Sulfoxide DMSO GC, Fisher Chemical™
Fisher Chemical
Dimethyl Sulfoxide DMSO GC, Fisher Chemical™
Dimethyl Sulfoxide DMSO GC Headspace 1L | CAS: 67-68-5 | C2H6OS | 78.13 g/mol
Have Questions?
Catalog number FCMD139-1
Price (MYR)
960.00
EA
Add to cart
Price (MYR)
960.00
EA
Add to cart
Dimethyl Sulfoxide DMSO GC Headspace 1L
  • Dimethyl Sulfoxide DMSO GC Headspace 1L Plastic Bottle Cas No. 67-68-5
Specifications
Chemical Name or MaterialDimethyl Sulfoxide
Melting Point18.4°C
CAS67-68-5
Boiling Point189°C
Quantity1 L
PackagingPlastic
Molecular FormulaC2H6OS
MDL NumberMFCD00002089
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
IUPAC Namemethanesulfinylmethane
Molecular Weight (g/mol)78.13
Unit SizeEA