Check Order Status
Barium Chloride Anhydrous (Laboratory), Fisher Chemical™
Element Search
Structure Search
Barium Chloride Anhydrous (Laboratory), Fisher Chemical™
Have Questions?
Change view
Quantity:
500 g
100 g
Packaging:
Poly Bottle
Catalog number
FCMB31-500
Price (MYR)
529.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
529.00
EA
Add to cart
Specifications
Boiling Point
1560°C
Chemical Name or Material
Barium Chloride Anhydrous
Melting Point
960°C
CAS
10361-37-2
ChemAlert Storage Symbol
Blue
View more