Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™
Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™
Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™
Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™
Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™
Fisher Chemical
Acetonitrile, Optima™, for HPLC and GC, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Catalog NumberQuantityPackaging
FCMA996-11 LWide Neck Amber Bottle
Catalog number FCMA996-1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Wide Neck Amber Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetonitrile
Cleanup SymbolSolvent
Melting Point-46°C
Absorbance0.005 AU max. at 254nm, 0.02 AU max. at 220nm, 0.04 AU max. at 210nm, 0.05 AU max. at 205nm, 0.07 AU max. at 200nm, 0.15 AU max. at 195nm, 1.00 AU max. at 190nm
CAS75-05-8
View more
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 190-254nm range, to reduce interferences for smoother baselines
  • Tested for HPLC gradient suitability
  • Tested to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only