Check Order Status
Acetonitrile, Optima UHPLC/MS Grade
Element Search
Structure Search
Fisher Chemical
Acetonitrile, Optima UHPLC/MS Grade
Have Questions?
Catalog number
FCMA956-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
Melting Point
-46°C
CAS
75-05-8
Color
Colorless
Boiling Point
81°C
Quantity
1 L
View more