Phenol (Liquid/Certified ACS), Fisher Chemical™
Phenol (Liquid/Certified ACS), Fisher Chemical™
Phenol (Liquid/Certified ACS), Fisher Chemical™
Phenol (Liquid/Certified ACS), Fisher Chemical™
Phenol (Liquid/Certified ACS), Fisher Chemical™
Have Questions?
Catalog NumberQuantityPackaging
FCMA931I-500500 mLGlass Bottle
Catalog number FCMA931I-500
Price (MYR)
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPhenol
CAS108-95-2
ChemAlert Storage SymbolWhite
Health Hazard 2DANGER!
Health Hazard 3Emergency Overview
Combustible liquid. Toxic by inhalation. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.
NFPA
Health:4
Flammability:2
Instability:0
View more