Ammonium Acetate (Crystalline/HPLC), Fisher Chemical
Ammonium Acetate (Crystalline/HPLC), Fisher Chemical
Ammonium Acetate (Crystalline/HPLC), Fisher Chemical
Ammonium Acetate (Crystalline/HPLC), Fisher Chemical
Fisher Chemical
Ammonium Acetate (Crystalline/HPLC), Fisher Chemical
Have Questions?
Catalog NumberQuantityPackaging
FCMA639-500500 gPoly Bottle
Catalog number FCMA639-500
Price (MYR)
530.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
530.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium Acetate
Melting Point114°C
CAS631-61-8
ChemAlert Storage SymbolGray