Check Order Status
1-Propanol (Certified), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
1-Propanol (Certified), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMA414-4
4 L
Glass Bottle
Catalog number
FCMA414-4
Price (MYR)
942.00
Bottle
Add to cart
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
942.00
Bottle
Add to cart
Specifications
Chemical Name or Material
1-Propanol
pH
7
Cleanup Symbol
Solvent
Melting Point
-127°C
CAS
71-23-8
View more