Check Order Status
Acetic Acid, Glacial (Certified ACS), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Acetic Acid, Glacial (Certified ACS), Fisher Chemical™
Have Questions?
Catalog Number
Quantity
Packaging
FCMA38S-500
500 mL
Safe-Cote™ Glass
Catalog number
FCMA38S-500
Price (MYR)
500.00
Each
Add to cart
Quantity:
500 mL
Packaging:
Safe-Cote™ Glass
Request bulk or custom format
Price (MYR)
500.00
Each
Add to cart
Specifications
Chemical Name or Material
Acetic Acid, Glacial
Cleanup Symbol
Acid
pH
2.5
CAS
64-19-7
ChemAlert Storage Symbol
Red
View more