Check Order Status
Aerosol™ OT, Anhydrous Solid, Fisher Chemical
Element Search
Structure Search
Aerosol™ OT, Anhydrous Solid, Fisher Chemical
Specific gravity 1.1
Have Questions?
Catalog Number
Quantity
FCMA349-500
500 g
Catalog number
FCMA349-500
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or Material
Aerosol™ OT, Solid Anhydrous
Color
White
CAS
577-11-7
ChemAlert Storage Symbol
Gray
Health Hazard 1
Signal Word: WARNING!
View more
Anionic waxy solid surfactant for applications where water cannot be tolerated
Efficient wetting agent, dispersant, and emulsifier for a variety of applications
Used to reduce surface tension and to incrase absorbency and penetration
This material is a waxy solid
Fisher Chemical also offers a 75% solution in ethanol and water (Catalog No.: A345)