Check Order Status
Sulfuric Acid (ACS Grade), Fisher Chemical™
Element Search
Structure Search
Fisher Chemical
Sulfuric Acid (ACS Grade), Fisher Chemical™
Approx. Normality is 36
Have Questions?
Catalog Number
Quantity
Packaging
FCMA300-212
2.5 L
Poly Bottle
Catalog number
FCMA300-212
Price (MYR)
1,023.00
Each
Add to cart
Quantity:
2.5 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
1,023.00
Each
Add to cart
Specifications
Chemical Name or Material
Sulfuric Acid
pH
0.3
Cleanup Symbol
Acid
Color
Colorless to Brown
CAS
7664-93-9
View more