Check Order Status
o-Phosphoric Acid, 85% (HPLC), Fisher Chemical
Element Search
Structure Search
Fisher Chemical
o-Phosphoric Acid, 85% (HPLC), Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FCMA260-500
500 mL
Flint Glass Bottle
Catalog number
FCMA260-500
Price (MYR)
476.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Flint Glass Bottle
Request bulk or custom format
Price (MYR)
476.00
BOT
Add to cart
Specifications
Chemical Name or Material
o-Phosphoric Acid
pH
1
Cleanup Symbol
Acid
Name Note
85%
CAS
7664-38-2
View more