OPTIZYME™ HaeIII (BsuRI), Fisher BioReagents™
Product Image
OPTIZYME™ HaeIII (BsuRI), Fisher BioReagents™
Have Questions?
Catalog NumberQuantity
FBRBP8002-12500 U
Catalog number FBRBP8002-1
Price (MYR)
-
Quantity:
2500 U
5'...G G^C C...3'
3'...C C^G G...5'
Conditions for 100% Activity:
  • 1X OPTIZYME Buffer 5: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2 , 100mM KCl and 0.1mg/ml BSA.
  • Incubate at 37°C.
Supplied with: 10X OPTIZYME Buffer 5
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 – 20%
  • Buffer 2: 0 - 20%
  • Buffer 3: 0 - 20%
  • Buffer 4: 50 - 100%
  • Buffer 5: 100%
Storage Buffer:
10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/ml BSA and 50% (v/v) glycerol.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with HaeIII.
Methylation Effects:
Dam: never overlaps - no effect.
Dcm: may overlap - no effect.
CpG: may overlap - no effect.
EcoKI: never overlaps - no effect.
EcoBI: never overlaps - no effect.
Specifications
Concentration10 U/μL
Components25 to 50% Water, >50% Glycerin
Incubator Temperature37°C
For Use With (Application)>95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with HaeIII
Storage Buffer10mM Tris HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Quantity2500 U
Cut SiteGG.CC
Product TypeHaeIII (BsuRI)
FormLiquid
Unit SizeEA
Contents & Storage
Keep container tightly closed