Phosphate Buffered Saline (PBS), 10X Powder, pH 7.4, Fisher BioReagents™
Phosphate Buffered Saline (PBS), 10X Powder, pH 7.4, Fisher BioReagents™
Actual product may vary
Phosphate Buffered Saline (PBS), 10X Powder, pH 7.4, Fisher BioReagents™
Phosphate Buffered Saline (PBS), 10X Powder, pH 7.4, Fisher BioReagents™
Fisher Chemical
Phosphate Buffered Saline (PBS), 10X Powder, pH 7.4, Fisher BioReagents™
Phosphate Buffered Saline (PBS), 10X Powder provides a convenient way to make 1 L of 10X stock PBS solution.
Have Questions?
Catalog NumberQuantity
FBRBP665-12 x 1 L
Catalog number FBRBP665-1
Price (MYR)
448.00
EA
Add to cart
Quantity:
2 x 1 L
Price (MYR)
448.00
EA
Add to cart
  • Provides a convenient way to make 1 L of 10X stock PBS solution
  • Eliminates the need to weigh and mix individual components
  • When reconstituted in ultrapure water such as BP2819, each packet yields 1 L of a 10X PBS solution containing 1.37M NaCl, 0.027M KCl, and 0.119M phosphates
  • Tested for the absence of DNase, RNase, and protease
  • PBS is a standard buffer used in many biochemical applications to maintain pH
  • Intended for laboratory research use only
Specifications
ColorWhite
Packaging TypeFoil Pack
Quantity2 x 1 L
FormSolid
Unit SizeEA
Contents & Storage
Room temperature