Check Order Status
2,2,2-Trifluoroethanol (Peptide Synthesis), Fisher BioReagents
Element Search
Structure Search
2,2,2-Trifluoroethanol (Peptide Synthesis), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP622-100
100 mL
Glass Bottle
Catalog number
FBRBP622-100
Price (MYR)
-
Quantity:
100 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Alpha Vector
Trifluoroethanol
Chemical Name or Material
2, 2, 2-Trifluoroethanol
Cleanup Symbol
Solvent
CAS
75-89-8
ChemAlert Storage Symbol
Red
View more
Trifluoroethanol is often used in peptide synthesis procedures.