Check Order Status
Lysozyme, Egg White (White Cryst. Powder)
Actual product may vary
Lysozyme, Egg White (White Cryst. Powder)
Have Questions?
Catalog Number
Quantity
FBRBP535-1
1 g
Catalog number
FBRBP535-1
Price (MYR)
-
Quantity:
1 g
Application:
This enzyme is used to lyse
E. coli
for preparation of plasmid DNA from transformed cell cultures
Not on TSCA inventory:
for research and development use only; not for manufacturing or commercial purposes
Enzyme Activity: ≈20,000U/mg dry weight
Solubility: Readily soluble at 5mg/mL in 0.1M KH
2
PO
4
to give colorless solution
Specifications
Color
White
For Use With (Application)
Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
Source
Egg white
Quantity
1 g
Product Type
Lysozyme
Form
Crystalline Powder
Unit Size
EA
Contents & Storage
<0°C