Check Order Status
n-Butanol (Clear, Colorless Liq.), Fisher BioReagents
Element Search
Structure Search
n-Butanol (Clear, Colorless Liq.), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP505-500
500 mL
Glass Bottle
Catalog number
FBRBP505-500
Price (MYR)
-
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
n-Butanol
Melting Point
-89°C
CAS
71-36-3
ChemAlert Storage Symbol
Red
Color
Colorless
View more
n-Butanol is used as an analytical reagent and solvent.