Check Order Status
Potassium Chloride (White Crystals), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Potassium Chloride (White Crystals), Fisher BioReagents
Have Questions?
Change view
Quantity:
1 kg
500 g
Packaging:
Poly Bottle
Catalog number
FBRBP366-1
Price (MYR)
644.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
644.00
Each
Add to cart
Specifications
Boiling Point
1420°C
Chemical Name or Material
Potassium Chloride
Melting Point
770°C
CAS
7447-40-7
ChemAlert Storage Symbol
Gray
View more
Potassium Chloride is used in buffers for molecular biology and cell culture applications.