Check Order Status
Potassium Phosphate Dibasic (Fine White Crystalline Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Potassium Phosphate Dibasic (Fine White Crystalline Powder), Fisher BioReagents™
Have Questions?
Change view
Quantity:
1 kg
500 g
Packaging:
View all
Poly Bottle
Catalog number
FBRBP363-1
Price (MYR)
564.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
564.00
Each
Add to cart
Specifications
Chemical Name or Material
Potassium Phosphate Dibasic
Melting Point
340°C
CAS
11-4-7758
ChemAlert Storage Symbol
Gray
Color
White
View more
Potassium Phosphate Dibasic (Fine White Crystalline Powder)
This potassium salt is used in buffers for molecular biology and cell culture applications.