Check Order Status
Sodium Chloride, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Chloride, Fisher BioReagents™
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP358-1
1 kg
Poly Bottle
Catalog number
FBRBP358-1
Price (MYR)
464.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
464.00
Each
Add to cart
Specifications
Boiling Point
1461°C
Chemical Name or Material
Sodium Chloride
Melting Point
801°C
CAS
7647-14-5
ChemAlert Storage Symbol
Gray
View more
Sodium Chloride
Sodium Chloride is a main component of many electrophoresis and molecular biology buffers and media.