Check Order Status
Polysorbate 20, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Polysorbate 20, Fisher BioReagents™
Non-ionic detergent | CAS: 9005-64-5 | (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | 522.68 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP337-500
500 mL
HDPE Bottle
Catalog number
FBRBP337-500
Price (MYR)
546.00
Bottle
Add to cart
Quantity:
500 mL
Packaging:
HDPE Bottle
Request bulk or custom format
Price (MYR)
546.00
Bottle
Add to cart
Chemical Identifiers
CAS
9005-64-5
Specifications
Chemical Name or Material
Polysorbate 20
Color
Amber
CAS
9005-64-5
ChemAlert Storage Symbol
Gray
Physical Form
Viscous Liquid
View more
Non-ionic detergent
This nonionic detergent is used in the isolation of water-soluble membrane proteins.