Sodium Citrate Dihydrate (Small, Colorless Granules), Fisher BioReagents™
Sodium Citrate Dihydrate (Small, Colorless Granules), Fisher BioReagents™
Sodium Citrate Dihydrate (Small, Colorless Granules), Fisher BioReagents™
Sodium Citrate Dihydrate (Small, Colorless Granules), Fisher BioReagents™
Fisher Chemical
Sodium Citrate Dihydrate (Small, Colorless Granules), Fisher BioReagents™
CAS: 6132-04-3 | C6H9Na3O9 | 294.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Poly Bottle
Catalog number FBRBP327-1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium Citrate Dihydrate
CAS6132-04-3
ChemAlert Storage SymbolGray
Health Hazard 2CAUTION!
CAS68-04-2
View more
Sodium Citrate Dihydrate (Small, Colorless Granules)
Sodium Citrate is used primarily in the preparation of SSC buffer used in Southern transfers.