Check Order Status
Ammonium Acetate (White Crystals), Fisher BioReagents
Element Search
Structure Search
Ammonium Acetate (White Crystals), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP326-500
500 g
Poly Bottle
Catalog number
FBRBP326-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ammonium Acetate
Melting Point
114°C
CAS
631-61-8
ChemAlert Storage Symbol
Gray
Color
White
View more
Ammonium Acetate can be used to precipitate DNA from enzymatic reactions.