Bis-Tris, Fisher BioReagents
Bis-Tris, Fisher BioReagents
Bis-Tris, Fisher BioReagents
Bis-Tris, Fisher BioReagents
Fisher Chemical
Bis-Tris, Fisher BioReagents
Commonly used buffering agent
Have Questions?
Catalog NumberQuantity
FBRBP301-100100 g
Catalog number FBRBP301-100
Price (MYR)
749.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
749.00
EA
Add to cart
Specifications
Chemical Name or MaterialBis-Tris
CAS6976-37-0
ChemAlert Storage SymbolGray
Melting Point103°C
ColorWhite
View more
This biological buffer has a usable pH range of 5.8 to 7.2.