Check Order Status
Pepstatin A, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Pepstatin A, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP2671-10
10 mg
Amber Glass
Catalog number
FBRBP2671-10
Price (MYR)
1,084.00
EA
Add to cart
Quantity:
10 mg
Packaging:
Amber Glass
Request bulk or custom format
Price (MYR)
1,084.00
EA
Add to cart
Specifications
Chemical Name or Material
Pepstatin A
CAS
26305-03-3
ChemAlert Storage Symbol
Gray
Melting Point
235°C
Color
White
View more
Pepstatin A is an inhibitor for pepsin, renin, cathepsin D, and other acid proteases.