Check Order Status
Miconazole (Free Base), Fisher BioReagents
Element Search
Structure Search
Miconazole (Free Base), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP2668-5
5 g
Amber Glass
Catalog number
FBRBP2668-5
Price (MYR)
-
Quantity:
5 g
Packaging:
Amber Glass
Request bulk or custom format
Specifications
Chemical Name or Material
Miconazole
Type
Miconazole
Name Note
Free Base
Physical Form
Solid
CAS
22916-47-8
View more
Miconazole is used as an antifungal agent.