Check Order Status
Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™
Broad spectrum antibiotic
Have Questions?
Change view
Quantity:
5 g
1 g
Packaging:
Amber Glass
Catalog number
FBRBP2653-5
Price (MYR)
1,056.00
EA
Add to cart
Quantity:
5 g
Packaging:
Amber Glass
Request bulk or custom format
Price (MYR)
1,056.00
EA
Add to cart
Specifications
Chemical Name or Material
Doxycycline Hydrochloride
Type
Doxycycline Hydrochloride
Physical Form
Powder/Solid
Absorbance
Inclusive between 300 at 349nm
CAS
10592-13-9
View more
Broad spectrum antibiotic
Broad spectrum antibiotic. Broad spectrum inhibitor of matrix metalloproteinases in vivo.