Check Order Status
Carbenicillin (Disodium Salt), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Carbenicillin (Disodium Salt), Fisher BioReagents
Have Questions?
Change view
Quantity:
5 g
250 mg
1 g
Packaging:
Amber Glass
Catalog number
FBRBP2648-5
Price (MYR)
969.00
EA
Add to cart
Quantity:
5 g
Packaging:
Amber Glass
Request bulk or custom format
Price (MYR)
969.00
EA
Add to cart
Specifications
Chemical Name or Material
Carbenicillin
Type
Carbenicillin
Name Note
Disodium Salt
Physical Form
Solid
CAS
4800-94-6
View more
Used for the selection of ampicillin-resistant transformed cells and for the study of penicillin-sensitive transpeptidases in cell wall biosynthesis. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria, Pseudomonas.