Amphotericin B (Yellow Solution/250μg/mL), Fisher BioReagents
Amphotericin B (Yellow Solution/250μg/mL), Fisher BioReagents
Amphotericin B (Yellow Solution/250μg/mL), Fisher BioReagents
Fisher Chemical
Amphotericin B (Yellow Solution/250μg/mL), Fisher BioReagents
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
20 mL
Packaging:
Poly Bottle
Catalog number FBRBP2645-50
Price (MYR)
874.00
Each
Quantity:
50 mL
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
874.00
Each
Specifications
Chemical Name or MaterialAmphotericin B
TypeAmphotericin B
Name Note250μg/mL
Physical FormLiquid
CAS1397-89-3
View more
Amphotericin B is effective against yeast and fungi. An antibiotic solution used in cell biology.