Mineral Oil, Fisher BioReagents
Mineral Oil, Fisher BioReagents
Actual product may vary
Mineral Oil, Fisher BioReagents
CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FBRBP2629-11 LGlass Bottle
Catalog number FBRBP2629-1
Price (MYR)
-
Quantity:
1 L
Packaging:
Glass Bottle
Glass Bottle; 1L
Mineral Oil can be used in PCR applications.
Specifications
Chemical Name or MaterialMineral Oil
Name NoteClear, Viscous Liquid
CAS8042-47-5
ChemAlert Storage SymbolGray
Health Hazard 2CAUTION!
Health Hazard 3Emergency Overview
May cause eye, skin, and respiratory tract irritation. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:0
Flammability:1
Instability:0
ColorWhite
Recommended StorageRT
Boiling Point260°C
PackagingGlass Bottle
Molecular FormulaC16H10N2Na2O7S2
Quantity1 L
Specific Gravity0.818 to 0.880 (at 25°C)
Synonymacid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
InChI KeyAEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILESC1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
IUPAC Namedisodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
Molecular Weight (g/mol)452.363
PubChem CID9566064
Vapor Pressure<0.1 mmHg
Physical FormLiquid
Unit SizeEA