Check Order Status
Heparin, Fisher BioReagents
Element Search
Structure Search
Heparin, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP2524-10
10 mg
Poly Vial
Catalog number
FBRBP2524-10
Price (MYR)
-
Quantity:
10 mg
Packaging:
Poly Vial
Request bulk or custom format
Specifications
Chemical Name or Material
Heparin
Name Note
From Porcine Intestinal Mucosa
CAS
9041-08-1
ChemAlert Storage Symbol
Gray
Health Hazard 2
CAUTION!
View more
Heparin is used as an anticoagulant in biochemical research.