Check Order Status
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents
Have Questions?
Change view
Quantity:
500 mL
100 mL
1 L
Packaging:
View all
Poly Bottle
Catalog number
FBRBP2482-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ethylenediamine Tetraacetic Acid
Color
Colorless
Name Note
0.5 M Solution, pH 8.0 (for DNA Work)
CAS
60-00-4
ChemAlert Storage Symbol
Gray
View more
EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work.